siRNA Details
Gene Symbol: FOXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207305.4 | NP_997188.2 | 1 | 761 | ||
GenBank mRNA | AY344640.1 | - | 1 | 549 | ||
BC089432.1 | - | 1 | 764 | 30555309 | ||
BC103886.2 | - | 1 | 462 | |||
BC136570.1 | - | 1 | 496 | 9020560 | ||
BC136571.1 | - | 1 | 496 | 9020561 | ||
U13223.1 | - | 1 | 169 |
Gene Symbol: FOXC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001453.2 | NP_001444.2 | 1 | 385 | ||
GenBank mRNA | BC134421.1 | - | 1 | 389 | ||
L12143.1 | - | 1 | 184 | |||
U13221.1 | - | 1 | 169 |
Gene Symbol: FOXC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005251.2 | NP_005242.1 | 1 | 452 | ||
GenBank mRNA | BC113437.1 | - | 1 | 452 | 8322489 | |
BC113439.1 | - | 1 | 452 | 8322491 |
Gene Symbol: FOXC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW271272.1 | - | 2 | 268 |
Gene Symbol: FOXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004472.2 | NP_004463.1 | 1 | 691 | ||
GenBank mRNA | U13222.1 | - | 1 | 169 | ||
U59832.1 | - | 1 | 691 |
Gene Symbol: FOXD4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012184.4 | NP_036316.1 | 1 | 645 | ||
GenBank mRNA | AF452723.1 | - | 1 | 493 | ||
AK122950.1 | - | 1 | 645 | |||
AK125398.1 | - | 1 | 1064 | |||
AY344639.1 | - | 1 | 557 | |||
BC136428.1 | - | 1 | 637 | 9020417 |
Gene Symbol: FOXD4L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199135.4 | NP_954586.4 | 1 | 560 | ||
GenBank mRNA | AY344641.1 | - | 1 | 560 | ||
AY344642.1 | - | 1 | 560 |
Gene Symbol: FOXD4L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199244.3 | NP_954714.2 | 1 | 475 |
Gene Symbol: FOXD4L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126334.1 | NP_001119806.1 | 1 | 1307 |
Gene Symbol: FOXD4L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085476.1 | NP_001078945.1 | 1 | 1067 | ||
GenBank mRNA | AF343006.1 | - | 1 | 112 | ||
BC103887.2 | - | 1 | 475 | 40004959 | ||
BC137365.1 | - | 1 | 555 | 9021363 | ||
BC137366.1 | - | 1 | 555 | 9021364 |
Gene Symbol: FOXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005250.2 | NP_005241.1 | 1 | 513 | ||
GenBank mRNA | BC100027.1 | - | 1 | 513 | 30920793 | |
BC117226.1 | - | 1 | 341 | 40125777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
