siRNA Details
Gene Symbol: PPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300981.1 | NP_001287910.1 | 5 | 512 | ||
NM_021130.4 | NP_066953.1 | 4 | 355 | |||
GenBank mRNA | AB451307.1 | - | 4 | 266 | ||
AB451438.1 | - | 4 | 266 | |||
AK026569.1 | - | 1 | 310 | |||
AK130101.1 | - | 4 | 311 | |||
AK290085.1 | - | 4 | 311 | |||
AK290851.1 | - | 4 | 313 | |||
AK293003.1 | - | 5 | 468 | |||
AK316564.1 | - | 4 | 310 | |||
BC000689.2 | - | 1 | 288 | 3349335 | ||
BC003026.1 | - | 1 | 294 | 2823490 | ||
BC005320.1 | - | 1 | 310 | 3935025 | ||
BC007104.1 | - | 1 | 311 | 4281376 | ||
BC013915.1 | - | 1 | 315 | 3684721 | ||
BC018843.2 | - | 1 | 345 | |||
BC073992.1 | - | 1 | 300 | 6161389 | ||
BC093076.1 | - | 5 | 377 | 30414589 | ||
BC106030.1 | - | 1 | 302 | 2986147 | ||
BC137057.1 | - | 4 | 355 | 9021054 | ||
BC137058.1 | - | 4 | 355 | 9021055 | ||
BM806224.1 | - | 5 | 366 | |||
BM909285.1 | - | 32 | ||||
BU786475.1 | - | 4 | 349 | |||
CD014106.1 | - | 5 | 468 | |||
CR456707.1 | - | 4 | 266 | |||
EU794650.1 | - | 1 | 302 | |||
Y00052.1 | - | 4 | 280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
