siRNA Details
Gene Symbol: KRTAP19-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181607.1 | NP_853638.1 | 1 | 243 | ||
GenBank mRNA | AB096943.1 | - | 1 | 275 | ||
AJ457067.1 | - | 1 | 239 | |||
BC126284.1 | - | 1 | 304 | 8992000 | ||
BC126286.1 | - | 1 | 304 | 8992002 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
