siRNA Details
Gene Symbol: GUSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003504.3 | 3, 4 | 389 | |||
GenBank mRNA | BC034017.1 | - | 3, 4 | 390 | ||
BC067351.1 | - | 1, 2 | 87 | |||
BC140728.1 | - | 2, 3 | 212 | |||
BC171739.1 | - | 2, 3 | 212 | 9052954 |
Gene Symbol: GUSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027386.2 | 3, 4 | 367 | |||
GenBank mRNA | AK289965.1 | - | 2, 3 | 339 | ||
BT006760.1 | - | 1, 2 | 42 | |||
DT218991.1 | - | 3, 4 | 287 |
Gene Symbol: GUSBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033968.1 | 1, 2 | 99 | |||
GenBank mRNA | AW500063.1 | - | 1, 2 | 32 | ||
BC104473.1 | - | 1, 2 | 99 | |||
CF141910.1 | - | 1, 2 | 105 |
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029426.1 | 3, 4 | 285 | |||
GenBank mRNA | AK094021.1 | - | 4, 5 | 691 | ||
AK097091.1 | - | 4, 5 | 665 | |||
AK289832.1 | - | 3, 4 | 367 | |||
AK315849.1 | - | 3, 4 | 358 | |||
BC131740.1 | - | 1, 2 | 99 | |||
X83300.1 | - | 1, 2 | 108 |
Gene Symbol: SMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034021.1 | 4, 5 | 361 | |||
GenBank mRNA | AK289851.1 | - | 3, 4 | 287 | ||
CN312642.1 | - | 1, 2 | 151 | |||
X83301.1 | - | 4, 5 | 361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
