siRNA Details
Gene Symbol: GUSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003504.3 | 3 | 361 | |||
GenBank mRNA | BC034017.1 | - | 3 | 362 | ||
BC067351.1 | - | 1 | 59 | |||
BC140728.1 | - | 2 | 184 | |||
BC171739.1 | - | 2 | 184 | 9052954 |
Gene Symbol: GUSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027386.2 | 3 | 339 | |||
GenBank mRNA | AK289965.1 | - | 2 | 311 | ||
BT006760.1 | - | 1 | 14 | |||
DT218991.1 | - | 3 | 259 | |||
X83299.1 | - | 3 | 290 |
Gene Symbol: GUSBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033968.1 | 1 | 71 | |||
GenBank mRNA | BC104473.1 | - | 1 | 71 | ||
CF141910.1 | - | 1 | 77 |
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029426.1 | 3 | 257 | |||
GenBank mRNA | AK094021.1 | - | 4 | 663 | ||
AK097091.1 | - | 4 | 637 | |||
AK289832.1 | - | 3 | 339 | |||
AK315849.1 | - | 3 | 330 | |||
BC131740.1 | - | 1 | 71 | |||
X83300.1 | - | 1 | 80 |
Gene Symbol: SMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034021.1 | 4 | 333 | |||
GenBank mRNA | AK289851.1 | - | 3 | 259 | ||
CN312642.1 | - | 1 | 123 | |||
X83301.1 | - | 4 | 333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
