siRNA Details
Gene Symbol: RPLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003.2 | NP_000994.1 | 2 | 232 | ||
GenBank mRNA | AK026579.1 | - | 2 | 232 | ||
AK130958.1 | - | 2 | 235 | |||
AK311456.1 | - | 2 | 498 | |||
AY303789.1 | - | 2 | 233 | |||
BC003369.1 | - | 2 | 215 | 2900846 | ||
BC007590.1 | - | 2 | 203 | 3343021 | ||
CR450339.1 | - | 2 | 103 | |||
CR542209.1 | - | 2 | 103 | |||
M17886.1 | - | 2 | 232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
