siRNA Details
Gene Symbol: GNG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017998.3 | NP_001017998.1 | 1 | 117 | ||
NM_001198664.1 | NP_001185593.1 | 1 | 117 | |||
GenBank mRNA | AF493877.1 | - | 1 | 53 | ||
BC010384.1 | - | 1 | 79 | |||
BC015206.2 | - | 1 | 63 | |||
BC016319.1 | - | 1 | 75 | |||
BC072671.1 | - | 1 | 68 | 5768882 | ||
BC107574.1 | - | 1 | 84 | 4050245 | ||
BG708408.1 | - | 1 | 122 | |||
DB504443.1 | - | 1 | 117 | |||
U31383.1 | - | 1 | 76 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
