siRNA Details
Gene Symbol: KRT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005557.3 | NP_005548.2 | 5, 6 | 1191 | ||
GenBank mRNA | AF061812.1 | - | 5, 6 | 1066 | ||
AK290853.1 | - | 5, 6 | 1129 | |||
BC039169.1 | - | 5, 6 | 1102 | 4752995 | ||
S79867.1 | - | 5, 6 | 1052 |
Gene Symbol: KRT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000526.4 | NP_000517.2 | 5, 6 | 1107 | ||
GenBank mRNA | BC002690.2 | - | 5, 6 | 1092 | 3610489 | |
BC019097.2 | - | 5, 6 | 1098 | 5013623 | ||
BC042437.1 | - | 5, 6 | 1062 | 4776763 | ||
BC094830.1 | - | 5, 6 | 1098 | 5443153 | ||
BT007186.1 | - | 5, 6 | 1046 |
Gene Symbol: KRT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002275.3 | NP_002266.2 | 5, 6 | 1210 | ||
XM_011524784.2 | XP_011523086.1 | 6, 7 | 2814 | |||
XM_017024614.1 | XP_016880103.1 | 5, 6 | 1277 | |||
GenBank mRNA | AK090604.1 | - | 7, 8 | 1763 | ||
AK091239.1 | - | 5, 6 | 662 | |||
AK122864.1 | - | 5, 6 | 4605 | |||
AL832226.1 | - | 4, 5 | 3128 | |||
BC002641.2 | - | 5, 6 | 1075 | 3610392 | ||
BT007261.1 | - | 5, 6 | 1019 | |||
X07696.1 | - | 5, 6 | 1080 |
Gene Symbol: KRT17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000422.2 | NP_000413.1 | 5, 6 | 1074 | ||
GenBank mRNA | AK095342.1 | - | 5, 6 | 1003 | ||
BC000159.2 | - | 5, 6 | 1003 | 2986100 | ||
BC011901.2 | - | 5, 6 | 1001 | 4111703 | ||
BC056421.1 | - | 5, 6 | 1003 | 5208304 | ||
BC072018.1 | - | 6, 7 | 941 | 3531173 | ||
BC072019.1 | - | 5, 6 | 999 | 5442690 | ||
BX647923.1 | - | 4, 5 | 1340 | |||
EF608070.1 | - | 1 | 38 | |||
X05803.1 | - | 4, 5 | 455 | |||
X62571.1 | - | 5, 6 | 1001 |
Gene Symbol: KRT17P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK296148.1 | - | 5, 6 | 1099 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
