siRNA Details
Gene Symbol: KRT17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000422.2 | NP_000413.1 | 4 | 939 | ||
GenBank mRNA | AK095342.1 | - | 4 | 868 | ||
BC000159.2 | - | 4 | 868 | 2986100 | ||
BC011901.2 | - | 4 | 866 | 4111703 | ||
BC056421.1 | - | 4 | 868 | 5208304 | ||
BC072018.1 | - | 5 | 806 | 3531173 | ||
BC072019.1 | - | 4 | 864 | 5442690 | ||
BX647923.1 | - | 3 | 1205 | |||
X05803.1 | - | 3 | 320 | |||
X62571.1 | - | 4 | 866 |
Gene Symbol: KRT17P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK296148.1 | - | 4 | 964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
