siRNA Details
Gene Symbol: LINC00152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024204.1 | 4 | 752 | |||
NR_024205.1 | 3 | 588 | ||||
NR_024206.1 | 2 | 442 | ||||
GenBank mRNA | BC009508.2 | - | 2 | 407 | ||
BC070202.1 | - | 365 | ||||
BM989746.1 | - | 2 | 437 | |||
CF129206.1 | - | 2 | 480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
