siRNA Details
Gene Symbol: MIR4435-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC070201.1 | - | 2 | 327 |
Gene Symbol: LINC00152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024204.1 | 4 | 664 | |||
NR_024205.1 | 3 | 500 | ||||
NR_024206.1 | 2 | 354 | ||||
GenBank mRNA | AK312010.1 | - | 3 | 413 | ||
BC009508.2 | - | 2 | 319 | |||
BC070202.1 | - | 277 | ||||
BM989746.1 | - | 2 | 349 | |||
CF129206.1 | - | 2 | 392 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
