siRNA Details
Gene Symbol: IFNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172138.1 | NP_742150.1 | 2 | 89 | ||
GenBank mRNA | AY129148.1 | - | 2 | 89 | ||
AY184373.1 | - | 1 | 25 | |||
AY336715.1 | - | 1 | 28 | |||
BC113581.1 | - | 2 | 92 | 8322633 | ||
BC113583.1 | - | 2 | 92 | 8322635 | ||
DQ126337.1 | - | 2 | 89 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
