siRNA Details
Gene Symbol: COMMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284377.1 | NP_001271306.1 | 5 | 313 | ||
NM_001284378.1 | NP_001271307.1 | 5 | 313 | |||
NM_001284379.1 | NP_001271308.1 | 4 | 291 | |||
NM_001321844.1 | NP_001308773.1 | 5 | 307 | |||
NM_001321845.1 | NP_001308774.1 | 3 | 175 | |||
NM_001321846.1 | NP_001308775.1 | 4 | 247 | |||
NM_001321847.1 | NP_001308776.1 | 4 | 247 | |||
NM_001321848.1 | NP_001308777.1 | 4 | 291 | |||
NM_017828.4 | NP_060298.2 | 5 | 313 | |||
NR_104312.1 | 5 | 313 | ||||
XM_005254509.3 | XP_005254566.1 | 5 | 339 | |||
XM_005254510.3 | XP_005254567.1 | 4 | 273 | |||
XM_011521739.1 | XP_011520041.1 | 5 | 348 | |||
XM_011521740.1 | XP_011520042.1 | 4 | 282 | |||
XM_011521741.1 | XP_011520043.1 | 5 | 348 | |||
XM_017022385.1 | XP_016877874.1 | 4 | 282 | |||
GenBank mRNA | AK000459.1 | - | 5 | 250 | ||
AK057145.1 | - | 2 | 990 | |||
AK225716.1 | - | 5 | 274 | |||
AK293708.1 | - | 4 | 548 | |||
AK314740.1 | - | 5 | 246 | |||
AL136872.1 | - | 5 | 233 | |||
AY542160.1 | - | 4 | 219 | |||
BC000837.2 | - | 4 | 216 | |||
BC033744.1 | - | 2 | 1009 | |||
BC068998.1 | - | 5 | 235 | 6586373 | ||
BI829264.1 | - | 5 | 289 | |||
BI829484.1 | - | 5 | 313 | |||
BQ233598.1 | - | 276 | ||||
CR533452.1 | - | 4 | 219 | |||
HY162014.1 | - | 4 | 292 |
Gene Symbol: GOLGA6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022406.1 | XP_016877895.1 | 4 | 749 |
Gene Symbol: LOC732265 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022776.1 | XP_016878265.1 | 5 | 661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
