siRNA Details
Gene Symbol: SUDS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022491.2 | NP_071936.2 | 7 | 707 | ||
XM_011538671.2 | XP_011536973.1 | 8 | 1784 | |||
XM_017019817.1 | XP_016875306.1 | 6 | 591 | |||
XM_017019818.1 | XP_016875307.1 | 9 | 939 | |||
XM_017019819.1 | XP_016875308.1 | 8 | 801 | |||
XM_017019820.1 | XP_016875309.1 | 9 | 833 | |||
GenBank mRNA | AK023801.1 | - | 2 | 183 | ||
AK024460.1 | - | 7 | 690 | |||
BC030252.1 | - | 1 | 80 | |||
BC093990.1 | - | 2 | 137 | 7939835 | ||
BC098404.1 | - | 7 | 730 | 30332266 | ||
BC112000.1 | - | 2 | 137 | |||
BX647687.1 | - | 8 | 665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
