siRNA Details
Gene Symbol: EP400NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090610.1 | - | 4 | 1726 | ||
AK091234.1 | - | 3 | 1281 | |||
AK097720.1 | - | 4 | 846 | |||
AK126212.1 | - | 3 | 1402 | |||
AK289710.1 | - | 5 | 1357 | |||
AL117637.1 | - | 1 | 981 | |||
BC066974.1 | - | 5 | 1280 |
Gene Symbol: EP400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015409.4 | NP_056224.3 | 2 | 1351 | ||
GenBank mRNA | AB040931.1 | - | 1 | 1204 | ||
AK096311.1 | - | 2 | 1311 | |||
AY044869.1 | - | 1 | 1235 | |||
BC037208.1 | - | 2 | 1300 | |||
BC064554.1 | - | 2 | 1308 | |||
BC150291.1 | - | 1 | 1204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
