siRNA Details
Gene Symbol: AQP7P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015366.1 | XP_016870855.1 | 5 | 614 | ||
XM_017015367.1 | XP_016870856.1 | 4 | 531 | |||
XM_017015368.1 | XP_016870857.1 | 4 | 514 | |||
XM_017015369.1 | XP_016870858.1 | 5 | 597 | |||
XM_017015370.1 | XP_016870859.1 | 2 | 752 | |||
XM_017015371.1 | XP_016870860.1 | 3 | 381 | |||
XM_017015372.1 | XP_016870861.1 | 2 | 752 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
