siRNA Details
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170.2 | NP_001161.1 | 7 | 1047 | ||
NM_001318156.1 | NP_001305085.1 | 6 | 929 | |||
NM_001318157.1 | NP_001305086.1 | 7 | 1365 | |||
NM_001318158.1 | NP_001305087.1 | 7 | 1047 | |||
NR_134513.1 | 5 | 804 | ||||
NR_134514.1 | 6 | 923 | ||||
NR_134515.1 | 7 | 1074 | ||||
XM_005251453.3 | XP_005251510.1 | 8 | 1147 | |||
XM_006716765.3 | XP_006716828.1 | 6 | 808 | |||
XM_011517866.2 | XP_011516168.1 | 7 | 957 | |||
XM_011517867.2 | XP_011516169.1 | 5 | 587 | |||
XM_017014700.1 | XP_016870189.1 | 7 | 1248 | |||
XM_017014701.1 | XP_016870190.1 | 6 | 918 | |||
XM_017014702.1 | XP_016870191.1 | 7 | 953 | |||
XM_017014703.1 | XP_016870192.1 | 6 | 706 | |||
XM_017014704.1 | XP_016870193.1 | 5 | 1014 | |||
XM_017014705.1 | XP_016870194.1 | 5 | 684 | |||
XM_017014706.1 | XP_016870195.1 | 8 | 1078 | |||
GenBank mRNA | AB006190.1 | - | 7 | 884 | ||
AK055744.1 | - | 1 | 631 | |||
AK294028.1 | - | 7 | 858 | |||
AK297860.1 | - | 7 | 952 | |||
AK297885.1 | - | 7 | 1181 | |||
AK300716.1 | - | 4 | 825 | |||
AK301949.1 | - | 5 | 767 | |||
AK310456.1 | - | 5 | 786 | |||
AK315879.1 | - | 4 | 816 | |||
BC062701.1 | - | 6 | 756 | 30324263 | ||
BC119672.1 | - | 6 | 755 | |||
BC119673.1 | - | 6 | 755 | |||
DQ786216.1 | - | 2 | 259 |
Gene Symbol: AQP7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002817.2 | 4 | 682 | |||
GenBank mRNA | AK128231.1 | - | 4 | 678 | ||
BC070322.1 | - | 3 | 422 |
Gene Symbol: AQP7P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015366.1 | XP_016870855.1 | 8 | 1147 | ||
XM_017015367.1 | XP_016870856.1 | 7 | 1064 | |||
XM_017015368.1 | XP_016870857.1 | 7 | 1047 | |||
XM_017015369.1 | XP_016870858.1 | 8 | 1130 | |||
XM_017015370.1 | XP_016870859.1 | 4 | 1147 | |||
XM_017015371.1 | XP_016870860.1 | 6 | 914 | |||
XM_017015372.1 | XP_016870861.1 | 5 | 1285 | |||
XM_017015373.1 | XP_016870862.1 | 5 | 785 | |||
XM_017015374.1 | XP_016870863.1 | 5 | 723 | |||
XM_017015375.1 | XP_016870864.1 | 4 | 639 |
Gene Symbol: LOC100509620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005276965.4 | XP_005277022.1 | 4 | 538 | ||
XM_006712884.3 | XP_006712947.1 | 4 | 962 | |||
XM_006712887.3 | XP_006712950.1 | 6 | 915 | |||
XM_017005467.1 | XP_016860956.1 | 3 | 534 | |||
XM_017005468.1 | XP_016860957.1 | 3 | 442 | |||
XM_017005469.1 | XP_016860958.1 | 3 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
