siRNA Details
Gene Symbol: LOC100509620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005276965.4 | XP_005277022.1 | 5 | 643 | ||
XM_006712884.3 | XP_006712947.1 | 5 | 1067 | |||
XM_006712887.3 | XP_006712950.1 | 7 | 1020 | |||
XM_017005467.1 | XP_016860956.1 | 4 | 639 | |||
XM_017005468.1 | XP_016860957.1 | 4 | 547 | |||
XM_017005469.1 | XP_016860958.1 | 4 | 536 |
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170.2 | NP_001161.1 | 8 | 1152 | ||
NM_001318156.1 | NP_001305085.1 | 7 | 975 | |||
NM_001318157.1 | NP_001305086.1 | 7 | 1828 | |||
NM_001318158.1 | NP_001305087.1 | 7 | 1510 | |||
NR_134513.1 | 5 | 1267 | ||||
NR_134514.1 | 6 | 1386 | ||||
NR_134515.1 | 7 | 1537 | ||||
XM_005251453.3 | XP_005251510.1 | 9 | 1252 | |||
XM_006716765.3 | XP_006716828.1 | 7 | 913 | |||
XM_011517866.2 | XP_011516168.1 | 8 | 1062 | |||
XM_011517867.2 | XP_011516169.1 | 6 | 692 | |||
XM_017014700.1 | XP_016870189.1 | 8 | 1353 | |||
XM_017014701.1 | XP_016870190.1 | 7 | 1023 | |||
XM_017014702.1 | XP_016870191.1 | 8 | 1058 | |||
XM_017014703.1 | XP_016870192.1 | 7 | 811 | |||
XM_017014704.1 | XP_016870193.1 | 6 | 1119 | |||
XM_017014705.1 | XP_016870194.1 | 6 | 789 | |||
XM_017014706.1 | XP_016870195.1 | 8 | 1541 | |||
GenBank mRNA | AB006190.1 | - | 8 | 989 | ||
AK055744.1 | - | 1 | 1094 | |||
BC062701.1 | - | 7 | 802 | 30324263 | ||
BC119672.1 | - | 6 | 1218 | |||
BC119673.1 | - | 6 | 1218 | |||
DQ786216.1 | - | 3 | 364 |
Gene Symbol: AQP7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002817.2 | 4 | 1145 | |||
GenBank mRNA | AK128231.1 | - | 4 | 1141 | ||
BC070322.1 | - | 4 | 468 |
Gene Symbol: AQP7P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015366.1 | XP_016870855.1 | 9 | 1252 | ||
XM_017015367.1 | XP_016870856.1 | 8 | 1169 | |||
XM_017015368.1 | XP_016870857.1 | 8 | 1152 | |||
XM_017015369.1 | XP_016870858.1 | 9 | 1235 | |||
XM_017015370.1 | XP_016870859.1 | 5 | 1252 | |||
XM_017015371.1 | XP_016870860.1 | 7 | 1019 | |||
XM_017015372.1 | XP_016870861.1 | 6 | 1331 | |||
XM_017015373.1 | XP_016870862.1 | 6 | 890 | |||
XM_017015374.1 | XP_016870863.1 | 6 | 828 | |||
XM_017015375.1 | XP_016870864.1 | 5 | 744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
