siRNA Details
Gene Symbol: KGFLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003674.2 | 3 | 766 | |||
NR_047527.1 | 4 | 876 | ||||
GenBank mRNA | AF523265.1 | - | 3 | 861 | ||
AK054997.1 | - | 2 | 1592 | |||
AK307215.1 | - | 3 | 497 | |||
AY098593.1 | - | 3 | 757 | |||
DB501751.1 | - | 4 | 417 |
Gene Symbol: FGF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002009.3 | NP_002000.1 | 4 | 1023 | ||
GenBank mRNA | AK313160.1 | - | 4 | 941 | ||
CR542002.1 | - | 3 | 494 | |||
M60828.1 | - | 4 | 939 | |||
S81661.1 | - | 4 | 939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
