siRNA Details
Gene Symbol: RPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000981.3 | NP_000972.1 | 2 | 91 | ||
XM_005257564.3 | XP_005257621.1 | 2 | 440 | |||
GenBank mRNA | AK130962.1 | - | 2 | 58 | ||
AK223082.1 | - | 2 | 53 | |||
AK311858.1 | - | 2 | 59 | |||
BC000530.2 | - | 2 | 41 | 2961653 | ||
BC013016.2 | - | 1 | 38 | 3010178 | ||
BC062709.1 | - | 2 | 56 | 6457109 | ||
BC066315.1 | - | 2 | 57 | 5287470 | ||
BC095445.1 | - | 2 | 91 | 30721753 | ||
BX537435.1 | - | 2 | 56 | |||
S56985.1 | - | 1 | 40 | |||
X63527.1 | - | 2 | 57 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
