siRNA Details
Gene Symbol: OR4M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004719.2 | NP_001004719.2 | 1 | 286 | ||
GenBank mRNA | BC136985.1 | - | 1 | 286 | 9020977 |
Gene Symbol: OR4M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005500.1 | NP_001005500.1 | 1 | 188 | ||
XM_017021338.1 | XP_016876827.1 | 1 | 847 | |||
XM_017021339.1 | XP_016876828.1 | 1 | 847 | |||
GenBank mRNA | BC136987.1 | - | 1 | 269 | 9020979 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
