siRNA Details
Gene Symbol: OR4M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004719.2 | NP_001004719.2 | 1 | 346 | ||
GenBank mRNA | BC136985.1 | - | 1 | 346 | 9020977 |
Gene Symbol: LOC102723532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546201.1 | XP_011544503.1 | 7 | 651 | ||
XM_011546205.1 | XP_011544507.1 | 11 | 1062 | |||
XM_011546206.1 | XP_011544508.1 | 10 | 1039 | |||
XM_017030114.1 | XP_016885603.1 | 2 | 93 | |||
XM_017030118.1 | XP_016885607.1 | 2 | 58 |
Gene Symbol: LOC727924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015416.1 | 10 | 1280 | |||
GenBank mRNA | BC048276.1 | - | 10 | 1280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
