siRNA Details
Gene Symbol: POM121L8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024583.1 | 1 | 1635 | |||
GenBank mRNA | AK302811.1 | - | 1 | 1638 | ||
AK308294.1 | - | 1 | 585 |
Gene Symbol: LOC102724728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136575.1 | 1 | 1563 | |||
GenBank mRNA | AK302597.1 | - | 1 | 1563 |
Gene Symbol: LOC727983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC112340.1 | - | 1 | 1300 |
Gene Symbol: POM121L10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024593.1 | 1 | 1563 |
Gene Symbol: POM121L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024591.1 | 1 | 1635 | |||
GenBank mRNA | AK292412.1 | - | 1 | 1567 |
Gene Symbol: POM121L9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003714.1 | 1 | 1563 | |||
GenBank mRNA | AL117401.1 | - | 1 | 1357 | ||
AY358961.1 | - | 1 | 1606 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
