siRNA Details
Gene Symbol: RAB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204883.1 | NP_001191812.1 | 3 | 414 | ||
NM_001204884.1 | NP_001191813.1 | 3 | 402 | |||
NM_001204885.1 | NP_001191814.1 | 3 | 392 | |||
NM_001204886.1 | NP_001191815.1 | 3 | 429 | |||
NM_001204887.1 | NP_001191816.1 | 2 | 680 | |||
NM_001204888.1 | NP_001191817.1 | 2 | 666 | |||
NM_198490.2 | NP_940892.1 | 2 | 680 | |||
GenBank mRNA | AK128345.1 | - | 2 | 476 | ||
AK291014.1 | - | 2 | 490 | |||
AY166852.2 | - | 2 | 658 | |||
BC062319.1 | - | 2 | 516 | 4873224 | ||
BX331971.2 | - | 2 | 667 | |||
D45418.1 | - | 2 | 469 | |||
DA008447.1 | - | 3 | 402 | |||
DA103180.1 | - | 3 | 392 | |||
DA317502.1 | - | 3 | 394 | |||
DA339537.1 | - | 3 | 429 | |||
DA757569.1 | - | 3 | 415 |
Gene Symbol: ISY1-RAB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204890.1 | NP_001191819.1 | 12 | 1267 | ||
GenBank mRNA | AB032986.1 | - | 12 | 1073 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
