siRNA Details
Gene Symbol: RAB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204883.1 | NP_001191812.1 | 4 | 651 | ||
NM_001204884.1 | NP_001191813.1 | 4 | 639 | |||
NM_001204885.1 | NP_001191814.1 | 4 | 629 | |||
NM_001204886.1 | NP_001191815.1 | 4 | 666 | |||
NM_001204887.1 | NP_001191816.1 | 4 | 1011 | |||
NM_001204888.1 | NP_001191817.1 | 3 | 903 | |||
NM_198490.2 | NP_940892.1 | 3 | 917 | |||
GenBank mRNA | AK128345.1 | - | 3 | 713 | ||
AK291014.1 | - | 3 | 727 | |||
AY166852.2 | - | 3 | 895 | |||
BC062319.1 | - | 3 | 753 | 4873224 | ||
D45418.1 | - | 3 | 706 |
Gene Symbol: ISY1-RAB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204890.1 | NP_001191819.1 | 13 | 1504 | ||
GenBank mRNA | AB032986.1 | - | 13 | 1310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
