siRNA Details
Gene Symbol: RAB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204883.1 | NP_001191812.1 | 3, 4 | 443 | ||
NM_001204884.1 | NP_001191813.1 | 3, 4 | 431 | |||
NM_001204885.1 | NP_001191814.1 | 3, 4 | 421 | |||
NM_001204886.1 | NP_001191815.1 | 3, 4 | 458 | |||
NM_001204888.1 | NP_001191817.1 | 2, 3 | 695 | |||
NM_198490.2 | NP_940892.1 | 2, 3 | 709 | |||
GenBank mRNA | AK128345.1 | - | 2, 3 | 505 | ||
AK291014.1 | - | 2, 3 | 519 | |||
AY166852.2 | - | 2, 3 | 687 | |||
BC062319.1 | - | 2, 3 | 545 | 4873224 | ||
D45418.1 | - | 2, 3 | 498 | |||
DA008447.1 | - | 3, 4 | 431 | |||
DA103180.1 | - | 3, 4 | 421 | |||
DA317502.1 | - | 3, 4 | 423 | |||
DA339537.1 | - | 3, 4 | 458 |
Gene Symbol: ISY1-RAB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204890.1 | NP_001191819.1 | 12, 13 | 1296 | ||
GenBank mRNA | AB032986.1 | - | 12, 13 | 1102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
