siRNA Details
Gene Symbol: GOLGA6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022406.1 | XP_016877895.1 | 3, 4 | 721 |
Gene Symbol: LOC440292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522301.2 | XP_011520603.1 | 3, 4 | 295 | ||
XM_017022799.1 | XP_016878288.1 | 3, 4 | 715 |
Gene Symbol: LOC732265 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022776.1 | XP_016878265.1 | 4, 5 | 633 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
