siRNA Details
Gene Symbol: FAM133B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040057.2 | NP_001035146.1 | 9, 10 | 713 | ||
NM_001288584.1 | NP_001275513.1 | 8, 9 | 698 | |||
NM_152789.3 | NP_690002.2 | 8, 9 | 614 | |||
NR_109929.1 | 8, 9 | 573 | ||||
XM_011516004.2 | XP_011514306.1 | 8, 9 | 877 | |||
XM_011516005.2 | XP_011514307.1 | 8, 9 | 614 | |||
GenBank mRNA | AK125498.1 | - | 7, 8 | 1042 | ||
AK313692.1 | - | 8, 9 | 698 | |||
AL833918.2 | - | 9, 10 | 713 | |||
BC017874.1 | - | 1 | 610 | |||
BC032461.1 | - | 8, 9 | 559 | 5197918 | ||
BC057771.1 | - | 9, 10 | 704 | 5301001 | ||
BC062706.1 | - | 8, 9 | 618 | |||
BC090868.1 | - | 8, 9 | 597 | 6190536 | ||
BX337761.2 | - | 8, 9 | 510 |
Gene Symbol: FAM133CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027508.1 | 1 | 1048 |
Gene Symbol: LOC100421561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC157872.1 | - | 1 | 571 | 9057091 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
