siRNA Details
Gene Symbol: FAM133DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034169.1 | 1 | 283 |
Gene Symbol: FAM133A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171109.1 | NP_001164580.1 | 4 | 714 | ||
NM_001171110.1 | NP_001164581.1 | 5 | 749 | |||
NM_001171111.1 | NP_001164582.1 | 3 | 625 | |||
NM_173698.2 | NP_775969.1 | 4 | 694 | |||
XM_006724641.3 | XP_006724704.1 | 3 | 726 | |||
XM_011530925.2 | XP_011529227.1 | 4 | 667 | |||
XM_011530926.2 | XP_011529228.1 | 4 | 505 | |||
XM_011530927.2 | XP_011529229.1 | 4 | 544 | |||
XM_011530928.1 | XP_011529230.1 | 4 | 478 | |||
XM_011530929.1 | XP_011529231.1 | 5 | 513 | |||
XM_011530931.2 | XP_011529233.1 | 3 | 680 | |||
XM_017029454.1 | XP_016884943.1 | 5 | 3353 | |||
XM_017029455.1 | XP_016884944.1 | 6 | 686 | |||
XM_017029456.1 | XP_016884945.1 | 4 | 621 | |||
GenBank mRNA | AK094978.1 | - | 4 | 501 | ||
BC113385.1 | - | 1 | 211 | 8322437 | ||
BC113387.1 | - | 1 | 211 | 8322439 | ||
DA297338.1 | - | 4 | 501 | |||
DA414074.1 | - | 4 | 524 | |||
DB084469.1 | - | 3 | 436 |
Gene Symbol: FAM133B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040057.2 | NP_001035146.1 | 4 | 390 | ||
NM_001288584.1 | NP_001275513.1 | 3 | 375 | |||
NM_152789.3 | NP_690002.2 | 3 | 291 | |||
NR_109929.1 | 3 | 291 | ||||
XM_011516004.2 | XP_011514306.1 | 3 | 554 | |||
XM_011516005.2 | XP_011514307.1 | 3 | 291 | |||
GenBank mRNA | AK125498.1 | - | 3 | 287 | ||
AK313692.1 | - | 3 | 375 | |||
AL833918.2 | - | 4 | 390 | |||
BC017874.1 | - | 1 | 287 | |||
BC032461.1 | - | 3 | 236 | 5197918 | ||
BC057771.1 | - | 4 | 381 | 5301001 | ||
BC062706.1 | - | 3 | 295 | |||
BC090868.1 | - | 3 | 274 | 6190536 |
Gene Symbol: FAM133CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027508.1 | 1 | 727 |
Gene Symbol: LOC100421561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC157872.1 | - | 1 | 248 | 9057091 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
