siRNA Details
Gene Symbol: HIST1H2AJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021066.2 | NP_066544.1 | 1 | 281 | ||
GenBank mRNA | BC066232.1 | - | 1 | 281 | ||
BC066233.1 | - | 1 | 281 | |||
BC066234.1 | - | 1 | 281 | |||
BC066235.1 | - | 1 | 281 | |||
BC066236.1 | - | 1 | 281 | |||
BC066237.1 | - | 1 | 281 | |||
BC133048.1 | - | 1 | 370 | 40147070 | ||
BC133050.1 | - | 1 | 370 | 40147072 |
Gene Symbol: HIST1H2AI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003509.2 | NP_003500.1 | 1 | 292 | ||
GenBank mRNA | AK311766.1 | - | 1 | 318 | ||
BC112254.1 | - | 1 | 370 | 8327722 | ||
BC112256.1 | - | 1 | 370 | 8327724 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
