siRNA Details
Gene Symbol: NPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148896.4 | NP_683694.1 | 1 | 194 | ||
GenBank mRNA | AB085942.1 | - | 1 | 25 | ||
AF525737.1 | - | 1 | 59 | |||
AF525738.1 | - | 1 | 59 | |||
BC126128.1 | - | 1 | 107 | 8991844 | ||
BC126485.1 | - | 1 | 107 | 8992201 | ||
BC146875.1 | - | 1 | 107 | 9056803 | ||
BC146890.1 | - | 1 | 107 | 9056818 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
