siRNA Details
Gene Symbol: ZNF544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320767.1 | NP_001307696.1 | 6 | 716 | ||
NM_001320769.1 | NP_001307698.1 | 6 | 758 | |||
NM_001320774.1 | NP_001307703.1 | 6 | 758 | |||
NM_001320780.1 | NP_001307709.1 | 6 | 681 | |||
NM_001320782.1 | NP_001307711.1 | 6 | 681 | |||
NM_014480.3 | NP_055295.2 | 6 | 681 | |||
XM_011526738.2 | XP_011525040.1 | 7 | 794 | |||
XM_017026599.1 | XP_016882088.1 | 7 | 794 | |||
GenBank mRNA | AF020591.1 | - | 5 | 429 | ||
AK291656.1 | - | 6 | 433 | |||
BC042177.1 | - | 5 | 425 | |||
BC067271.1 | - | 5 | 409 | 6199648 | ||
CB997427.1 | - | 6 | 693 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
