siRNA Details
Gene Symbol: KRTAP4-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032524.1 | NP_115913.1 | 1 | 324 | ||
GenBank mRNA | AJ406936.1 | - | 1 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
