siRNA Details
Gene Symbol: ANKRD30BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026916.1 | 6 | 743 | |||
GenBank mRNA | AF427490.1 | - | 4 | 364 | ||
BC126176.1 | - | 3 | 202 | |||
BC126178.1 | - | 3 | 202 |
Gene Symbol: ANKRD30BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098688.1 | - | 2 | 219 | ||
BC031237.1 | - | 2 | 176 | |||
BC043208.2 | - | 8 | 1758 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
