siRNA Details
Gene Symbol: ANKRD30BL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027019.1 | 7 | 1266 | |||
NR_027020.2 | 5 | 964 | ||||
GenBank mRNA | BC045801.1 | - | 5 | 964 | ||
BC057764.1 | - | 7 | 1269 |
Gene Symbol: ANKRD30BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026916.1 | 6 | 899 | |||
GenBank mRNA | AF427490.1 | - | 4 | 520 | ||
DQ179138.1 | - | 1 | 114 |
Gene Symbol: LOC644669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027417.1 | 5 | 446 | |||
GenBank mRNA | AK097631.1 | - | 5 | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
