siRNA Details
Gene Symbol: CRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311.4 | NP_001302.1 | 2, 3 | 158 | ||
GenBank mRNA | BC002738.2 | - | 1, 2 | 92 | 3631097 | |
BQ128075.1 | - | 2, 3 | 163 | |||
BT007181.1 | - | 1, 2 | 44 | |||
BU500642.1 | - | 1, 2 | 102 | |||
U09770.1 | - | 1, 2 | 108 | |||
U58630.1 | - | 1, 2 | 44 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
