siRNA Details
Gene Symbol: LCE3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178435.3 | NP_848522.1 | 2 | 57 | ||
XM_011509484.1 | XP_011507786.1 | 2 | 153 | |||
GenBank mRNA | BC131716.1 | - | 2 | 57 | 40123193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
