siRNA Details
Gene Symbol: LCE3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032563.1 | NP_115952.1 | 2 | 162 | ||
GenBank mRNA | AB048288.1 | - | 2 | 162 | ||
BC069291.1 | - | 2 | 155 | 7262717 | ||
BC101784.1 | - | 2 | 145 | 8069290 | ||
BC120935.1 | - | 1 | 105 | 40115032 | ||
BC120936.1 | - | 1 | 103 |
Gene Symbol: LCE3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178435.3 | NP_848522.1 | 2 | 142 | ||
XM_011509484.1 | XP_011507786.1 | 2 | 238 | |||
GenBank mRNA | BC131716.1 | - | 2 | 142 | 40123193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
