siRNA Details
Gene Symbol: TMA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF077202.1 | - | 3 | 135 | ||
AF161448.1 | - | 2 | 84 | |||
AK126594.1 | - | 3 | 97 | |||
AL712164.1 | - | 3 | 112 | |||
AY508979.1 | - | 1 | 118 | |||
AY794023.1 | - | 3 | 119 | |||
BC001102.2 | - | 1 | 107 | |||
BC080531.1 | - | 3 | 120 | 4875991 | ||
BU536461.1 | - | 3 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
