siRNA Details
Gene Symbol: KRTAP4-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031854.2 | NP_114060.1 | 1 | 124 | ||
GenBank mRNA | AJ406943.1 | - | 1 | 124 | ||
BC004180.2 | - | 1 | 104 | 2959821 | ||
BC004212.2 | - | 1 | 104 | 2959821 | ||
BC101159.1 | - | 1 | 116 | 40015747 | ||
BC101160.1 | - | 1 | 116 | 40015748 | ||
BC101161.1 | - | 1 | 116 | |||
BC101162.1 | - | 1 | 116 | 40015750 |
Gene Symbol: KRTAP4-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033060.2 | NP_149049.1 | 1 | 132 | ||
GenBank mRNA | AJ406942.1 | - | 1 | 132 | ||
BC065372.1 | - | 2 | 629 | |||
BC103845.1 | - | 1 | 112 | 40002744 | ||
BC103846.2 | - | 1 | 112 | 40002745 |
Gene Symbol: KRTAP4-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033062.3 | NP_149051.1 | 1 | 123 | ||
GenBank mRNA | AJ406934.1 | - | 1 | 123 | ||
BC127252.1 | - | 1 | 103 | 40030890 |
Gene Symbol: KRTAP4-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033187.1 | NP_149443.1 | 1 | 91 | ||
GenBank mRNA | AJ406935.1 | - | 1 | 85 |
Gene Symbol: KRTAP4-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033188.3 | NP_149445.3 | 1 | 114 | ||
GenBank mRNA | AJ406937.1 | - | 1 | 114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
