siRNA Details
Gene Symbol: KRTAP4-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033062.3 | NP_149051.1 | 1 | 423 | ||
GenBank mRNA | AJ406934.1 | - | 1 | 423 | ||
BC127252.1 | - | 1 | 403 | 40030890 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
