siRNA Details
Gene Symbol: KRTAP4-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031854.2 | NP_114060.1 | 1 | 61 | ||
GenBank mRNA | AJ406943.1 | - | 1 | 61 | ||
BC004180.2 | - | 1 | 41 | 2959821 | ||
BC004212.2 | - | 1 | 41 | 2959821 | ||
BC101159.1 | - | 1 | 53 | 40015747 | ||
BC101160.1 | - | 1 | 53 | 40015748 | ||
BC101161.1 | - | 1 | 53 | |||
BC101162.1 | - | 1 | 53 | 40015750 |
Gene Symbol: KRTAP4-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033062.3 | NP_149051.1 | 1 | 60 | ||
GenBank mRNA | AJ406934.1 | - | 1 | 60 | ||
BC127252.1 | - | 1 | 40 | 40030890 |
Gene Symbol: KRTAP4-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032524.1 | NP_115913.1 | 1 | 56 | ||
GenBank mRNA | AJ296168.1 | - | 1 | 59 | ||
AJ406936.1 | - | 1 | 56 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
