siRNA Details
Gene Symbol: RPL23AP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026981.1 | 4 | 847 | |||
NR_026982.1 | 3 | 775 | ||||
GenBank mRNA | AK293095.1 | - | 3 | 498 | ||
BC065556.1 | - | 3 | 642 |
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC065480.1 | - | 4 | 783 |
Gene Symbol: HLA-F-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026972.1 | 6 | 1678 | |||
NR_026973.1 | 2 | 462 | ||||
GenBank mRNA | AK092748.1 | - | 6 | 1678 | ||
BM726335.1 | - | 2 | 420 |
Gene Symbol: RPL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000984.5 | NP_000975.2 | 3 | 335 | ||
GenBank mRNA | AK130965.1 | - | 3 | 324 | ||
AK312123.1 | - | 3 | 324 | |||
BC014459.1 | - | 1 | 315 | 4872948 | ||
BC058041.1 | - | 3 | 327 | 6716166 | ||
BG476195.1 | - | 3 | 444 | |||
BP382007.1 | - | 3 | 335 | |||
L13799.1 | - | 2 | 285 | |||
U02032.1 | - | 3 | 313 | |||
U37230.1 | - | 1 | 334 | |||
U43701.1 | - | 3 | 332 |
Gene Symbol: RPL23AP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002229.2 | 1 | 435 | |||
GenBank mRNA | BC096706.1 | - | 1 | 435 | ||
BC096740.1 | - | 1 | 435 | |||
BC098172.1 | - | 1 | 435 |
Gene Symbol: RPL23AP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC146939.1 | - | 1 | 411 | 9056867 | |
BC146942.1 | - | 1 | 411 | 9056870 |
Gene Symbol: RPL23AP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003572.2 | 4 | 728 | |||
GenBank mRNA | AK127000.1 | - | 3 | 457 | ||
BC052779.1 | - | 5 | 788 | |||
BC064938.1 | - | 4 | 637 |
Gene Symbol: RPL23AP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_000029.3 | 3 | 532 | |||
NR_024528.1 | 4 | 783 | ||||
NR_024529.1 | 3 | 689 | ||||
GenBank mRNA | BC000596.1 | - | 3 | 483 | ||
BC105600.1 | - | 3 | 484 | |||
BX643196.1 | - | 3 | 505 |
Gene Symbol: RPL23AP87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029406.1 | 4 | 1685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
