siRNA Details
Gene Symbol: KRTAP19-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181611.1 | NP_853642.1 | 1 | 13 | ||
GenBank mRNA | AB096946.1 | - | 1 | 40 | ||
BC100836.1 | - | 1 | 13 | 40002081 | ||
BC100837.2 | - | 1 | 13 | 40002082 | ||
BC100838.2 | - | 1 | 13 | 40002083 | ||
BC100839.2 | - | 1 | 13 | 40002084 | ||
BC113020.1 | - | 1 | 13 | 40013750 | ||
BC113021.1 | - | 1 | 13 | 40013751 | ||
BC128032.1 | - | 1 | 13 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
