siRNA Details
Gene Symbol: SSU72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014188.2 | NP_054907.1 | 5 | 900 | ||
GenBank mRNA | AF161530.1 | - | 5 | 640 | ||
AF277178.1 | - | 5 | 579 | |||
AJ276409.1 | - | 5 | 688 | |||
AK001809.1 | - | 5 | 648 | |||
AK127149.1 | - | 3 | 1836 | |||
BC008070.2 | - | 5 | 848 | 2961422 | ||
BC018909.2 | - | 5 | 884 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
