siRNA Details
Gene Symbol: KRTAP19-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181609.3 | NP_853640.1 | 1 | 13 | ||
GenBank mRNA | AB096944.1 | - | 1 | 41 | ||
AF139541.1 | - | 1 | 13 | |||
BC126280.1 | - | 1 | 57 | |||
BC126282.1 | - | 1 | 57 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
