siRNA Details
Gene Symbol: KRTAP19-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181609.3 | NP_853640.1 | 1 | 36 | ||
GenBank mRNA | AB096944.1 | - | 1 | 64 | ||
AF139541.1 | - | 1 | 36 | |||
BC126280.1 | - | 1 | 80 | |||
BC126282.1 | - | 1 | 80 |
Gene Symbol: KRTAP19-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181610.1 | NP_853641.1 | 1 | 36 | ||
GenBank mRNA | AB096945.1 | - | 1 | 59 | ||
BC117197.1 | - | 1 | 80 | 40125748 | ||
BC117199.1 | - | 1 | 80 | 40125750 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
