siRNA Details
Gene Symbol: KRTAP4-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033060.2 | NP_149049.1 | 2 | 411 | ||
GenBank mRNA | AJ406942.1 | - | 2 | 411 | ||
BC065372.1 | - | 3 | 908 | |||
BC103845.1 | - | 2 | 391 | 40002744 | ||
BC103846.2 | - | 2 | 391 | 40002745 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
