siRNA Details
Gene Symbol: KRTAP17-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031964.1 | NP_114170.1 | 1 | 98 | ||
GenBank mRNA | AJ406952.1 | - | 1 | 98 | ||
BC069078.1 | - | 1 | 96 | |||
BC069297.1 | - | 1 | 98 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
