siRNA Details
Gene Symbol: PNRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017761.3 | NP_060231.1 | 3 | 1253 | ||
XM_017001691.1 | XP_016857180.1 | 3 | 1158 | |||
GenBank mRNA | AF374386.1 | - | 3 | 1172 | ||
AK000319.1 | - | 2 | 974 | |||
AK300522.1 | - | 3 | 994 | |||
AK311305.1 | - | 4 | 1038 | |||
BC001959.1 | - | 3 | 1220 | 3536977 | ||
BC078177.1 | - | 3 | 1200 | 6526331 | ||
BC085018.1 | - | 3 | 1223 | 6650445 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
